1,1-Dimethyl-3-hydroxypyrrolidinium Bromide 1 * 100 mg
Catalog # PROO-D456998-100MG
Supplier: LGC Standards PROMOCHEM
1,1-Dimethyl-3-hydroxypyrrolidinium Bromide 1 * 100 mg
Catalog # PROO-D456998-100MG
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Sulphuric acid 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1086804
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
VWR®, Colour standards, Platinum-Cobalt (Pt-Co, Hazen)
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
Supplier: VWR Chemicals
Lead acetate cotton Reag. Ph. Eur. 1048101
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
pH buffer solutions, colour coded
Supplier: Hach
pH Standard Solutions, IUPAC, Certified
Supplier: Hach