ContactUsLinkComponent
PhaseTwo VS4-3 Vapour Shipper Static Holding Time (days) - 23 Normal Evaporation Rate (liters/day) - 0.19 Liquid Nitrogen Capacity - 4.4 litres Neck Diameter (mm) - 71 Overall Height / Diameter (mm) - 467 x 234 Weight Empty / Full (kg) - 6 / 9.47 (Wa 1 * 1 items
Catalog # TAYW10-152-01-0037
Supplier:  TAYLOR WHARTON
PhaseTwo VS4-3 Vapour Shipper Static Holding Time (days) - 23 Normal Evaporation Rate (liters/day) - 0.19 Liquid Nitrogen Capacity - 4.4 litres Neck Diameter (mm) - 71 Overall Height / Diameter (mm) - 467 x 234 Weight Empty / Full (kg) - 6 / 9.47 (Wa 1 * 1 items
Catalog # TAYW10-152-01-0037
Supplier:  TAYLOR WHARTON
Frequently Bought Together