ContactUsLinkComponent
Volumetric flask, 75 ml, class A, conformity certified, NS12/21, NS hexagonal hollow stopper, Diffico brown, made of DURAN tube, all volumetric flasks are marked with batch number and an enclosed batch certificate per original packaging 1 * 1 items
Catalog # WITG3.668.0075
Supplier:  witeg Labortechnik
Volumetric flask, 75 ml, class A, conformity certified, NS12/21, NS hexagonal hollow stopper, Diffico brown, made of DURAN tube, all volumetric flasks are marked with batch number and an enclosed batch certificate per original packaging 1 * 1 items
Catalog # WITG3.668.0075
Supplier:  witeg Labortechnik
Frequently Bought Together