ContactUsLinkComponent
Hoshizaki UK Model No. ADVANCE F 140-4 C U Advance Freezer, reach-in, two-section, 1200 litre, -25 to -12 C temperature range, mono-block design, (1) right hand hinged door &(1) left hand hinged door, (8) grey shelves (2/1GN deep), LED lighting, AI 1 * 1 items
  HOSHADVANCEF1404CU
 :  HOSHIZAKI
Hoshizaki UK Model No. ADVANCE F 140-4 C U Advance Freezer, reach-in, two-section, 1200 litre, -25 to -12 C temperature range, mono-block design, (1) right hand hinged door &(1) left hand hinged door, (8) grey shelves (2/1GN deep), LED lighting, AI 1 * 1 items
  HOSHADVANCEF1404CU
 :  HOSHIZAKI
Frequently Bought Together