3-O-ALLYL ROCURONIUM BROMIDE >90% 1 * 25 mg
Catalog # PROOTRCP769550-25
Supplier: LGC Standards PROMOCHEM
3-O-ALLYL ROCURONIUM BROMIDE >90% 1 * 25 mg
Catalog # PROOTRCP769550-25
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets
Supplier: Mettler - Toledo
pH Standard Solutions, IUPAC, Certified
Supplier: Hach