Number of racks 6 Number of 2ml vials 6000 Boxes per rack 10 Static evaporation rate (L/day) 0.98 Static holding time (days) 173 LN2 capacity (L) 175 Outer diameter (mm) 673 Outer height(mm) 963 Neck opening (mm) 216 1 * 1 items
Catalog # TAYW10-152-01-0054
Supplier: TAYLOR WHARTON