ContactUsLinkComponent
Perfluorooctane sulfonic acid [CAS:1763-23-1] 100ug/ml in Methanol ,, CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a freezer at -18C or below 1 * 1 mL
Catalog # CMPLP253009
Perfluorooctane sulfonic acid [CAS:1763-23-1] 100ug/ml in Methanol ,, CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a freezer at -18C or below 1 * 1 mL
Catalog # CMPLP253009
Frequently Bought Together