POLYSORBATE 80 1 * 5 g
PHR3790-5X1G
: Merck
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, DuraCal
: HAMILTON BONADUZ
pH buffer solutions
: Mettler - Toledo
Crucibles, porcelain
: MORGAN ADVANCED MATERIALS HALD
Potassium bromate ≥99%
: Thermo Fisher Scientific