ContactUsLinkComponent
5 components: 1000mg/l each of Ca Mg K Fe Na in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
Catalog # CMPLRM038755L1
5 components: 1000mg/l each of Ca Mg K Fe Na in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
Catalog # CMPLRM038755L1
Frequently Bought Together