ContactUsLinkComponent
Markers, 35bp + 5000bp, 3.2mL. 35 bp lower marker 5,000 bp upper marker. For use with the Fragment Analyzer systems and the DNF-915 kit or the ZAG DNA Analyzer system and the ZAG-110 kit. 1 * 3,2 mL
Catalog # AGLSFS-SMK480-0003
Supplier:  AGILENT
Markers, 35bp + 5000bp, 3.2mL. 35 bp lower marker 5,000 bp upper marker. For use with the Fragment Analyzer systems and the DNF-915 kit or the ZAG DNA Analyzer system and the ZAG-110 kit. 1 * 3,2 mL
Catalog # AGLSFS-SMK480-0003
Supplier:  AGILENT