ContactUsLinkComponent
100bp DNA Ladder, 3.2mL. 100-1,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-910, DNF-935 kits or the ZAG DNA Analyzer system and the ZAG-135 kits 1 * 3 mL
Catalog # AGLSFS-SLR910-0003
Supplier:  AGILENT
100bp DNA Ladder, 3.2mL. 100-1,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-910, DNF-935 kits or the ZAG DNA Analyzer system and the ZAG-135 kits 1 * 3 mL
Catalog # AGLSFS-SLR910-0003
Supplier:  AGILENT