ContactUsLinkComponent
Microscope cover glasses, 18x50 mm, borosilicate glass D 263 M, thickness No. 1.5H (0.17±0.005 mm), CE marked, 100 pcs./hinged-lid plastic box 1000/carton 1 * 1.000 items
  MARI0107992094
Microscope cover glasses, 18x50 mm, borosilicate glass D 263 M, thickness No. 1.5H (0.17±0.005 mm), CE marked, 100 pcs./hinged-lid plastic box 1000/carton 1 * 1.000 items
  MARI0107992094
Frequently Bought Together