ContactUsLinkComponent
Protect Select - For preservation of Anaerobes with orange cryobeads in moisture resistant freezer box with user reference grid on box lid and inside base. Organisms can be frozen and recovered successfully at -20C, -70C and -80C. A full list of r 1 * 25 items
Catalog # TSCLTS/73-AN25
Supplier:  TSC LIMITED
Protect Select - For preservation of Anaerobes with orange cryobeads in moisture resistant freezer box with user reference grid on box lid and inside base. Organisms can be frozen and recovered successfully at -20C, -70C and -80C. A full list of r 1 * 25 items
Catalog # TSCLTS/73-AN25
Supplier:  TSC LIMITED
Frequently Bought Together