ContactUsLinkComponent
RETSCH Test Sieve DIN ISO 3310/1 8x1 315µm stainless steel with Calibration Certificate 1 * 1 items
Catalog # RETS60.340.000315
Supplier:  Retsch GmbH
RETSCH Test Sieve DIN ISO 3310/1 8x1 315µm stainless steel with Calibration Certificate 1 * 1 items
Catalog # RETS60.340.000315
Supplier:  Retsch GmbH
Frequently Bought Together