ContactUsLinkComponent
PhaseTwo ICE 44 freezer with controller Static Holding Time (days) - 18 Normal Evaporation Rate (liters/day) - 8 Liquid Nitrogen Capacity - 157 litres 2ml Vial Capacity - Up to 43,600 (Standard Config - 42,900) Neck Diameter (mm) - 457 Overall Height 1 * 1 items
  TAYW10-152-01-0060
PhaseTwo ICE 44 freezer with controller Static Holding Time (days) - 18 Normal Evaporation Rate (liters/day) - 8 Liquid Nitrogen Capacity - 157 litres 2ml Vial Capacity - Up to 43,600 (Standard Config - 42,900) Neck Diameter (mm) - 457 Overall Height 1 * 1 items
  TAYW10-152-01-0060
Frequently Bought Together