2,7-DIBROMO-9H-FLUOREN-9-ONE 1 * 25 g
Catalog # APOSOR22580-25G
Supplier: Apollo Scientific
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 124,000 99-100%, hydrolysed
Supplier: Thermo Fisher Scientific
Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific