UNIVERSAL ADAPTER 6V 5A 1 * 1 items
Catalog # OPTKALIM03
Supplier: OPTIKA
Frequently Bought Together
SWITCH ON/OFF 1 * 1 items
Supplier: ERLAB CAPTAIRE
HOSE NOZZLE DN 15 MM / G1/2INCH 1 * 1 items
Supplier: VACUUBRAND
CLASP COMPLETE 120V 1 * 1 items
Supplier: Hettich
Seal for filter adapter 1 * 5 items
Supplier: EPPENDORF
HANDLE OF LOCKING V2 FOR PRECELLYS 24 AN 1 * 1 items
Supplier: Bertin Instruments
Accessories for compact UV lamps
Supplier: UVP ULTRA VIOLET PRODUCTS
GASKET 1 * 1 items
Supplier: Thermo Scientific
Door contact switch (1S) 250VAC / 25W 1 * 1 items
Supplier: Binder
DRIVE BELT FOR PHMT 1 * 1 items
Supplier: GRANT INSTRUMENTS
CABLE MVP TO ML600 1 * 1 items
Supplier: HAMILTON BONADUZ