Ethyl 4-hydroxybenzoate, 99% 1 * 25 kg
A13172.PG
Frequently Bought Together
2-Butoxyethanol 99%, extra pure
: Thermo Fisher Scientific
Bromoform 96% stabilised
: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
n-Hexane, extra dry 96+%, AcroSeal®
: Thermo Fisher Scientific
n-Hexane 95% for residue analysis, for trace analysis of polyaromatic hydrocarbons
: Thermo Fisher Scientific
Methyl ethyl ketone ACS
: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
: Thermo Fisher Scientific
tert-Butanol ACS
: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% ACS
: Thermo Fisher Scientific
2-Methylbutane ≥99%, Reagent Grade
: Honeywell Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals