0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Volumetric flasks, USP, BLAUBRAND® ETERNA, class A, DE-M, 500 ml, NS 19/26, Boro 3.3, with PP stopper, USP individual certificate 1 * 1 items
Catalog # BRND956852
Supplier:
Brand
Expand image
Volumetric flasks, USP, BLAUBRAND® ETERNA, class A, DE-M, 500 ml, NS 19/26, Boro 3.3, with PP stopper, USP individual certificate 1 * 1 items
Catalog # BRND956852
Supplier:
Brand
Frequently Bought Together
Measuring cylinders, tall form, class A, borosilicate glass, BLAUBRAND®
Supplier: Brand
VWR®, Hydrometers, M100
Supplier: VWR Collection
BLAUBRAND® Volumetric Flasks, Class A, Blue Markings
Supplier: Brand
Bulb pipettes (one mark), BLAUBRAND®, class AS
Supplier: Brand
Automatic burettes, Dr. Schilling pattern, SILBERBRAND
Supplier: Brand
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
DURAN® Pasteur Pipettes
Supplier: DWK Life Sciences
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
Volumetric flasks, class A, amber markings, BLAUBRAND® ETERNA
Supplier: Brand
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection
Short hydrometers
Supplier: Alla France
VWR® Burettes, with PTFE Key Stopcock
Supplier: VWR Collection