Perform st.conc. OXY (50x) 40 g BT 1 * 2 kg
SCUE70003329
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
VWR® PureStep, Multi-Layer Adhesive Floor Mats
: VWR Collection
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Cleanroom vacuum cleaner, CWR POLY series
: TIGER-VAC
Instrument trays, with lid
: BOCHEM