Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SLIDE 76X26X1MM CUT TWIN-FROSTED ALU BA 1 * 2.500 items
Catalog # MARI1005200
Supplier:
MARIENFELD
Expand image
SLIDE 76X26X1MM CUT TWIN-FROSTED ALU BA 1 * 2.500 items
Catalog # MARI1005200
Supplier:
MARIENFELD
Frequently Bought Together
Push button safety blood collection sets, BD Vacutainer®
Supplier: BD Medical
SUPERFLO™ Histology Cassettes, One-Piece Moulded
Supplier: Simport Scientific
BD Plastipak™ Syringes
Supplier: BD Medical
Cassette, PM Embedding, 10 tubes x75 wi 1 * 750 items
Supplier: Epredia
Cassette, PM Embedding 1000/bx no lids 1 * 1.000 items
Supplier: Epredia
Slide mailers, UniMailer™
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Slimsette®, M511
Supplier: Simport Scientific