ContactUsLinkComponent
CAPACITY: 91 LITRES OPERATING TEMPERATURE: +4oC TEMPERATURE RANGE:+2oC to +10oC 2 SHELVES + 1 BIN AUTOMATIC DEFROST MOULDED PLASTIC INTERIOR, ENAMELLED STEEL EXTERIOR SOLID DOOR 2 ADJUSTABLE FEET AT FRONT INTERNAL LIGHT NOISE LEVEL: 39 dB(A) ENERGY C 1 * 1 items
Catalog # BORORLPL04044
Supplier:  Labcold
CAPACITY: 91 LITRES OPERATING TEMPERATURE: +4oC TEMPERATURE RANGE:+2oC to +10oC 2 SHELVES + 1 BIN AUTOMATIC DEFROST MOULDED PLASTIC INTERIOR, ENAMELLED STEEL EXTERIOR SOLID DOOR 2 ADJUSTABLE FEET AT FRONT INTERNAL LIGHT NOISE LEVEL: 39 dB(A) ENERGY C 1 * 1 items
Catalog # BORORLPL04044
Supplier:  Labcold
Frequently Bought Together