ContactUsLinkComponent
Congealing point thermometer, similar DIN, solid stem, 0+100:0,5C, white backed, gallium filled, 300x6,5mm, total immersion, top finish round, durable pigment, government verified with Verification Certificate with 3 test points 1 * 1 items
Catalog # AMARM247300-GA/VC
Supplier:  Amarell
Congealing point thermometer, similar DIN, solid stem, 0+100:0,5C, white backed, gallium filled, 300x6,5mm, total immersion, top finish round, durable pigment, government verified with Verification Certificate with 3 test points 1 * 1 items
Catalog # AMARM247300-GA/VC
Supplier:  Amarell