ContactUsLinkComponent
SURESTART webseal mat, clear, eva (ethylene-vinyl-acetat), for 96 round well plates, 8mm diameter, flat base, no slit, 1 * 5 items
Catalog # HYPE60180-M101
SURESTART webseal mat, clear, eva (ethylene-vinyl-acetat), for 96 round well plates, 8mm diameter, flat base, no slit, 1 * 5 items
Catalog # HYPE60180-M101
Frequently Bought Together