Sodium sulfate anhydrous EMPROVE ESSENTIAL Ph Eur,BP,ChP,USP,E 514 1 * 25 kg
Catalog # 1.28211.8026
Supplier: MERCK PRODUCTION CHEMICALS
Sodium sulfate anhydrous EMPROVE ESSENTIAL Ph Eur,BP,ChP,USP,E 514 1 * 25 kg
Catalog # 1.28211.8026
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Sodium chloride ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water, EMSURE® for analysis, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ammonium acetate ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium hydrogen carbonate 99.7-100.3%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals