2-chloro-3-(2.4.5-trichlorophenoxy)-1.4-dih ydronaphthalene-1. 4-dione 1 * 100 mg
MAYBJFD00950
2-chloro-3-(2.4.5-trichlorophenoxy)-1.4-dih ydronaphthalene-1. 4-dione 1 * 100 mg
MAYBJFD00950
Frequently Bought Together
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
: VWR Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Diethyl ether, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised
: VWR Chemicals
Methanol, SupraSolv® for GC-MS
: Merck
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
: Thermo Fisher Scientific
Methanol ≥99.8%, Ultrapure for HPLC
: Thermo Fisher Scientific
2-Propanol ≥99.85%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Water, GPR RECTAPUR®
: VWR Chemicals
Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
2-Propanol, extra dry over Molecular Sieve 99.5%, AcroSeal®
: Thermo Fisher Scientific
Toluene ≥99.3%, Laboratory Reagent
: Honeywell Chemicals
Petroleum spirit, 50…70 °C ≥90%, AnalaR NORMAPUR®
: VWR Chemicals
Water, PESTINORM® for headspace gas chromatography
: VWR Chemicals