ContactUsLinkComponent
Oxygen Sensor S210.V01 Range 0-25% Required power supply 24V= @160mA Accuracy < 2%FS Expected sensor lifetime: 5 years (guaranteed 2 years) Recommended maintenance: 6 months LCD 4,3 - 65K color with touch screen technology RS485 Interface Two progr 1 * 1 items
Catalog # TAYW10-089-01-0001
Supplier:  TAYLOR WHARTON
Oxygen Sensor S210.V01 Range 0-25% Required power supply 24V= @160mA Accuracy < 2%FS Expected sensor lifetime: 5 years (guaranteed 2 years) Recommended maintenance: 6 months LCD 4,3 - 65K color with touch screen technology RS485 Interface Two progr 1 * 1 items
Catalog # TAYW10-089-01-0001
Supplier:  TAYLOR WHARTON