£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
Iron fe-2 testkit lovibond 1 * 1 items
LOVI2418440
:
Lovibond
Iron fe-2 testkit lovibond 1 * 1 items
LOVI2418440
:
Lovibond
Frequently Bought Together
Phosphate buffered saline, hematology reagent, for diagnostics use 1 * 10 L
: Avantor
Acid Red 183 30% (dye content), Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
RUCAPARIB (FREE BASE) 1 * 10 mg
: ENZO LIFE SCIENCES
Nitrite VHR L, 500 ml
: Lovibond
TOLUENE D8 1000PPB IN METHANOL 1 * 5 Ampoul
: CUSTOM MADE CHEMICALS LAB
Buffer concentrate, citrate phosphate, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY