Sodium sulfate anhydrous EMPROVE ESSENTIAL Ph Eur,BP,ChP,USP,E 514 1 * 2,5 kg
Catalog # 1.28211.2501
Supplier: MERCK PRODUCTION CHEMICALS
Sodium sulfate anhydrous EMPROVE ESSENTIAL Ph Eur,BP,ChP,USP,E 514 1 * 2,5 kg
Catalog # 1.28211.2501
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Magnesium oxide heavy 98.0-100.5% Ph. Eur.
Supplier: VWR Chemicals
Sodium chloride 99.0-100.5% USP, Ph. Eur., BP
Supplier: VWR Chemicals
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Disodium oxalate ≥99.8%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium gluconate, TECHNICAL
Supplier: VWR Chemicals
Potassium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ammonium peroxodisulphate (APS) ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium sulphate, anhydrous 98.5-101.0%, fine powder USP, Ph. Eur.
Supplier: VWR Chemicals
Potassium nitrate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium iodide ≥99%
Supplier: Thermo Fisher Scientific