Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Ba @ 300 Se @ 130 Sn @ 60 Cu @ 30 Li @ 25 Sb @ 20 Mo @ 10 Tl @ 8 Ag @ 7 Ni @ 5 Co, Cr @ 3 V @ 1 µg/mL in 5% 1 * 100 mL
Catalog # PROOVHGICHUSP-TELI
Supplier:
LGC Standards PROMOCHEM
Expand image
Ba @ 300 Se @ 130 Sn @ 60 Cu @ 30 Li @ 25 Sb @ 20 Mo @ 10 Tl @ 8 Ag @ 7 Ni @ 5 Co, Cr @ 3 V @ 1 µg/mL in 5% 1 * 100 mL
Catalog # PROOVHGICHUSP-TELI
Supplier:
LGC Standards PROMOCHEM
Frequently Bought Together
pH buffers, for LAQUAtwin meters
Supplier: HORIBA
Supelco® Organic Reference Standard, Hexanal
Supplier: Merck
Organic reference standard, Neoxanthin, analytical standard, Supelco®
Supplier: Merck
Supelco® Organic Reference Standard, F.A.M.E. Mix, C20:1-C20:5 Unsaturates
Supplier: Merck
EPA 8270 Phenols mix, certified reference material, Supelco®
Supplier: Merck
Polystyrene, analytical standards for GPC
Supplier: Merck
MINOCYCLINE HYDROCHLORIDE 1 * 75 mg
Supplier: Merck
TRANEXAMIC ACID RELATED COMPOUND C 1 * 30 mg
Supplier: Merck
Supelco® Organic Reference Standard, PCB No 28, 52, 101, 138, 153, 180 Solution
Supplier: Merck
5,5-(DIPHENYL-D10) HYDANTOIN 1 * 5 mg
Supplier: LGC Standards PROMOCHEM
pH buffer solutions, colour coded
Supplier: Hach
Petrolatum, white
Supplier: USP