ContactUsLinkComponent
ADAM-MC, the automated fluorescence cell counter performs viability and cell counting measurements by using the PI(Propidium Iodide) staining method combined with advanced image analysis. 1 * 1 items
Catalog # NANOADAM-MC-UK
Supplier:  NANOENTEK
ADAM-MC, the automated fluorescence cell counter performs viability and cell counting measurements by using the PI(Propidium Iodide) staining method combined with advanced image analysis. 1 * 1 items
Catalog # NANOADAM-MC-UK
Supplier:  NANOENTEK
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.