0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Clevenger apparatus for essential oils, borosilicate glass 3.3, NS 29/32, stopcock PTFE NS 14,5, bore 2,5mm 1 * 1 items
Catalog # LENZ81004237
Supplier:
Lenz Laborglas GmbH & CO.KG
Expand image
Clevenger apparatus for essential oils, borosilicate glass 3.3, NS 29/32, stopcock PTFE NS 14,5, bore 2,5mm 1 * 1 items
Catalog # LENZ81004237
Supplier:
Lenz Laborglas GmbH & CO.KG
Frequently Bought Together
VWR®, Beakers, Low Form
Supplier: VWR Collection
VWR®, Pasteur Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Sodium sulphate, anhydrous, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
VWR®, Beakers, Tall Form
Supplier: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection