ContactUsLinkComponent
Copper Cu - 1000 mg/l in diluted HNO3 for ICP CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 30 mL
Catalog # ACSDC015.2NP.L03
Supplier:  CPACHEM
Copper Cu - 1000 mg/l in diluted HNO3 for ICP CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 30 mL
Catalog # ACSDC015.2NP.L03
Supplier:  CPACHEM
Frequently Bought Together