Bromide (from KBr) Standard: Br- @ 10000 µg/mL in H2O 1 * 500 mL
Catalog # PROOVHG-I1PBR-500
Supplier: LGC Standards PROMOCHEM
Bromide (from KBr) Standard: Br- @ 10000 µg/mL in H2O 1 * 500 mL
Catalog # PROOVHG-I1PBR-500
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach