Order Entry
Northern Ireland
ContactUsLinkComponent
Stainless Steel (SS) Hose Clamps, 4-3/32" x 6"; 10/Pk
Stainless Steel (SS) Hose Clamps, 4-3/32" x 6"; 10/Pk
Catalog # MFLX06403-24
undefined
Stainless Steel (SS) Hose Clamps, 4-3/32" x 6"; 10/Pk
Catalog # MFLX06403-24

About this item

Ideal for securely clamping tubes in place.

  • Resists corrosion

The band-type worm drive clamp features stainless steel strap and screw housing with a stainless steel screw.

  • Max. int. Ø:
    152,4 mm (6")
  • Description:
    Stainless Steel (SS) Hose Clamps, 4-3/32" x 6"; 10/Pk
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Pk:
    10
  • Max. tubing ext.Ø:
    152,4 mm (6")
  • Min. int.Ø:
    103,98125 mm (4,09375")
Frequently Bought Together