ContactUsLinkComponent
Slide-On Ge Micro Tip ATR 350 Multi-coated, conical shape germanium tip crystal 350 micron spherical finish allows gentle contact with sample and small footprint High precision easy alignment slide mount Minimum throughput at default aperture 1 * 1 items
Catalog # THEC840-194800
Supplier:  THERMO EC
Slide-On Ge Micro Tip ATR 350 Multi-coated, conical shape germanium tip crystal 350 micron spherical finish allows gentle contact with sample and small footprint High precision easy alignment slide mount Minimum throughput at default aperture 1 * 1 items
Catalog # THEC840-194800
Supplier:  THERMO EC
Frequently Bought Together