MD-Piano 1200 Resin bonded diamond grinding surface for fine grinding of materials HV 150 - 2000. Surface finish comparable to SiC-P aper grit 1200 1 * 1 items
Catalog # MSPP40800138
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together