ContactUsLinkComponent
Resin bonded diamond grinding surface for plane grinding of materials HV 150 - 2000. Surface finish comparable to SiC-Paper grit 500 . For magnetic fixation on MD-disc.250 mm (10 ) dia. 1 * 1 items
  MSPP40800134
Resin bonded diamond grinding surface for plane grinding of materials HV 150 - 2000. Surface finish comparable to SiC-Paper grit 500 . For magnetic fixation on MD-disc.250 mm (10 ) dia. 1 * 1 items
  MSPP40800134
Frequently Bought Together