2-(2-HYDROXY-5-METHYLPHENYL)BENZOTRIAZOL 1 * 100 mg
Catalog # CRM96697-100MG
Supplier: Merck
Frequently Bought Together
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® ARISTAR®, Multi-Element Calibration Standard IV in dil. Nitric Acid for ICP
Supplier: VWR Chemicals
VWR®, Refractive index Certified Reference Material at 20, 25 and 30 °C
Supplier: VWR Chemicals
Potassium dichromate, Certipur®, Supelco®
Supplier: Merck
Humidity standards
Supplier: ROTRONIC INSTRUMENTS
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
Supelco® PESTANAL® Organic Reference Standard, Molinate
Supplier: Merck
VWR®, Flash point reference standard, Pensky Martens
Supplier: VWR Chemicals
Buffer Solutions
Supplier: Thermo Fisher Scientific
VWR®, Sulphur, Certified Reference Materials, in odourless kerosene
Supplier: VWR Chemicals
pH buffers, for LAQUAtwin meters
Supplier: HORIBA
pH buffer solutions, colour coded
Supplier: Hach
Supelco® PESTANAL® Organic Reference Standard, Omethoate
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
ICP METAL STANDARD 31 COMPONENT 1 * 100 mL
Supplier: CPACHEM
VWR®, Turbidity standards, formazin
Supplier: VWR Chemicals