ContactUsLinkComponent
Elbow with small flange KF DN 25 and hose nozzle for tubing i.d. 19 mm, PP 1 * 1 items
Catalog # VACU20751803
Supplier:  VACUUBRAND
Elbow with small flange KF DN 25 and hose nozzle for tubing i.d. 19 mm, PP 1 * 1 items
Catalog # VACU20751803
Supplier:  VACUUBRAND
Frequently Bought Together