Autodrain. Automatically drains any water left in the chamber at the end of the cycle to the drain (Approx water temp is 100C, so the drain cooling option is recommended) Note: Autofill should also be considered in conjunction with this option. 1 * 1 items
Catalog # ASTLAAP018
Supplier: AIR 7 SEAS TRANSPORT LOGISTICS, INC
Frequently Bought Together