OZONE GENERATOR BMT 803N Max. system pressure: 2.0 bar g Fittings: 1/4 Swagelok Supply: 230 VAC, 155 W, 50/60 Hz Power control: pot entiometer Ozone production 8 g/h at 100 g/Nm / 20C 1 * 1 items
Catalog # MSPP803N.00049
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together