ContactUsLinkComponent
OZONE ANALYZER BMT 965 ST Concentration range: 300 g/Nm Pressure range: 3.0 bar abs Fittings: for 3x5 mm FEP (in & out) Supply: 100 - 240 VAC, 15 VA, 50/60 Hz 1 * 1 items
  MSPP965ST.00271
OZONE ANALYZER BMT 965 ST Concentration range: 300 g/Nm Pressure range: 3.0 bar abs Fittings: for 3x5 mm FEP (in & out) Supply: 100 - 240 VAC, 15 VA, 50/60 Hz 1 * 1 items
  MSPP965ST.00271
Frequently Bought Together