ContactUsLinkComponent
Plastic feeding tubes, 16ga x 38mm. 50 sterile pouches of 5. Polyurethane tube with rounded tip. For mouse oral gavage. Disposable. Designed to minimize trauma to esophagus. Large ID for viscous compounds. Straight tube wipes clean to prevent aspirat 1 * 250 items
Catalog # MSPPFTPU-16-38
Plastic feeding tubes, 16ga x 38mm. 50 sterile pouches of 5. Polyurethane tube with rounded tip. For mouse oral gavage. Disposable. Designed to minimize trauma to esophagus. Large ID for viscous compounds. Straight tube wipes clean to prevent aspirat 1 * 250 items
Catalog # MSPPFTPU-16-38
Frequently Bought Together