MANIFOLD, HORIZONTAL, 3 WIDE NEXGEN RAT 1800, REGULAR TEMPERATURE POLYCARBONATE PLASTIC, WITH SIMULATOR END CAPS NOTE - COMPONENTS A RE AUTOCLAVABLE AND CAN BE CHEMICALLY STERILIZED, PLEASE SEND ALLENTOWN ADDITIONAL INFORMATION 1 * 1 SET
Catalog # MSPP414202-20
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together