ContactUsLinkComponent
Pre-cleaned and certified (<35 ppb) 40 mL glass vials pretreated with phosphoric acid (20 uL of 85 wt% H3PO4). Cap includes Teflon-lined septum and dust cap. 72 vials per case. Please note that the typical lot size is 20 cases per lot due to manufact 1 * 72 items
Catalog # IONIHMI90090-01
Supplier:  IONICS
Pre-cleaned and certified (<35 ppb) 40 mL glass vials pretreated with phosphoric acid (20 uL of 85 wt% H3PO4). Cap includes Teflon-lined septum and dust cap. 72 vials per case. Please note that the typical lot size is 20 cases per lot due to manufact 1 * 72 items
Catalog # IONIHMI90090-01
Supplier:  IONICS
Frequently Bought Together