ContactUsLinkComponent
Ag, Al, B, Ba, Bi, Ca, Cd, Cr, Cu, Fe, In, Li, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn @ 500 µg/g in 75 cSt Hydrocarbon Oil 1 * 400 g
Catalog # PROOVHGV26500400G
Ag, Al, B, Ba, Bi, Ca, Cd, Cr, Cu, Fe, In, Li, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn @ 500 µg/g in 75 cSt Hydrocarbon Oil 1 * 400 g
Catalog # PROOVHGV26500400G
Frequently Bought Together