Orders LinkContactUsLinkComponent
ABLE C competent cells
ABLE C competent cells
ABLE C electroporation-competent cells, reduces the number of common cloning vectors 4-fold, 5×0,1 ml
Catalog # AGLS200161
Supplier:   AGILENT
ABLE C competent cells
Catalog # AGLS200161
Supplier:  AGILENT
ABLE C competent cells
ABLE C electroporation-competent cells, reduces the number of common cloning vectors 4-fold, 5×0,1 ml
Price
Inquire For Price
Log in for availability
1 * 5 items
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
ABLE Competent Cells for toxic clones reduce copy number four to ten fold compared to the XL1-Blue strain.
  • Reduces the copy number of ColE1-derived plasmids (such as pUC and pBluescript® plasmids) four-fold
  • Decreases level of cloned protein product to increase cell viability
  • Transformation efficiency of 5×10⁶ to 2×10⁷ transformants/µg
  • Also available in an electroporation-competent version
ABLE C Competent Cells for toxic clones enhance the probability of cloning DNA encoding toxic proteins. By reducing the copy number of ColE1-derived plasmids, the ABLE C strain decreases the levels of cloned gene products and enhances the probability of propagating a toxic clone.
The ABLE C Competent Cells provide a viable alternative to commonly used cloning vectors with high copy numbers. Positive clones observed on initial screening of lambda libraries can be excised or re-cloned into any convenient vector and can be introduced into ABLE C strains.
ABLE K Competent Cells for toxic clones enhance the probability of cloning DNA encoding toxic proteins. By reducing the copy number of ColE1-derived plasmids, the ABLE K strain decreases the levels of cloned gene products and enhances the probability of propagating a toxic clone.
ABLE K Competent Cells provide a viable alternative to commonly used cloning vectors with high copy numbers. Positive clones observed on initial screening of lambda libraries can be excised or re-cloned into any convenient vector and can be introduced into ABLE K strains.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
  • Description:
    ABLE C electroporation-competent cells, reduces the number of common cloning vectors 4-fold, 5×0,1 ml
  • Seq:
    02