ContactUsLinkComponent
Pentapharm® PH-M170/01-51/9000-100_0-ZZZ-250-R Color 51/9000 - 1-RG-Blue, crystal clear-000 Add. Treatment ZZZ - No additional treatment-00 Surface 100_0 - Glossy / Glossy-00 Thickness 250 µm Width 136 - 138 mm Length Max reel dimension 500 mm (diame 1 * 1 Roll
Catalog # MSPPP024851
Pentapharm® PH-M170/01-51/9000-100_0-ZZZ-250-R Color 51/9000 - 1-RG-Blue, crystal clear-000 Add. Treatment ZZZ - No additional treatment-00 Surface 100_0 - Glossy / Glossy-00 Thickness 250 µm Width 136 - 138 mm Length Max reel dimension 500 mm (diame 1 * 1 Roll
Catalog # MSPPP024851
Frequently Bought Together