BioCircuit MEA 24 plate, 16 microelectrodes per well including one large stimulation electrode per well, clear polystyrene walls wit h an opaque bottom, includes on-plate spotting guides, box of 5 plates 1 * 5 items
Catalog # MSPPM384-BIO-24-5
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together